Learn More
Cayman Chemical 5-(4'-(Azidomethyl)(1,1'-biphenyl)-2-yl)-2H-tetrazole, Purity: ≥95%, CAS: 152708-24-2, 10 mg
SDP
Catalog No. 502917042
Encompass_Preferred
A potential impurity found in commercial preparations of tetrazole-containing sartans
Catalog No. 50-291-7042 Supplier Cayman Chemical Supplier No. 3878010 MG
A potential impurity found in commercial preparations of tetrazole-containing sartans