Learn More
Medchemexpress LLC 5-(4-butoxy-3-chlorophenyl)-N-[(2-morpholin-4-ylpyridin-3-yl)methyl]pyridine-3-carboxamide | 1266212-81-0 | 99.1% | 480.99 g/mol | C26H29ClN4O3 | 1 ML
SDP
Catalog No. 5000215652
Encompass_Preferred
A-887826 is a potent, selective, voltage-dependent Nav1.8 sodium channel blocker (IC50 = 11 nM) used in research on pain mechanisms. It is orally bioavailable and reduces neuropathic tactile allodynia in vivo. Supplied as a 10 mM solution in DMSO, it is suitable for in vitro pharmacology, electrophysiology, and preclinical pain studies.
  • Potent Nav1.8 inhibition (IC50 = 11 nM).
  • Voltage-dependent mode of action.
  • Demonstrated efficacy in neuropathic pain models.
  • Supplied as a ready-to-use DMSO solution for convenient dosing.
  • Suitable for electrophysiology and pharmacology assays.
Catalog No. 50-002-15652 Supplier Medchemexpress LLC Supplier No. HY10008010MM 1ML
A-887826 is a potent, selective, voltage-dependent Nav1.8 sodium channel blocker (IC50 = 11 nM) used in research on pain mechanisms. It is orally bioavailable and reduces neuropathic tactile allodynia in vivo. Supplied as a 10 mM solution in DMSO, it is suitable for in vitro pharmacology, electrophysiology, and preclinical pain studies.
  • Potent Nav1.8 inhibition (IC50 = 11 nM).
  • Voltage-dependent mode of action.
  • Demonstrated efficacy in neuropathic pain models.
  • Supplied as a ready-to-use DMSO solution for convenient dosing.
  • Suitable for electrophysiology and pharmacology assays.