Learn More
Medchemexpress LLC 5-(4-butoxy-3-chlorophenyl)-N-((2-morpholinopyridin-3-yl)methyl)nicotinamide | 1266212-81-0 | >97.0% | 480.99 g/mol | C26H29ClN4O3 | 10 MG
SDP
Catalog No. 5000214989
Encompass_Preferred
A-887826 is a small-molecule, voltage-dependent NaV1.8 sodium channel blocker originally described by Abbott Laboratories. It is orally bioavailable, exhibits potent in vitro inhibition of NaV1.8 (IC50 ≈ 11 nM), and reduces neuropathic tactile allodynia in rodent models.
  • Potent NaV1.8 inhibition (IC50 ≈ 11 nM).
  • Orally bioavailable with demonstrated in vivo efficacy in neuropathic pain models.
  • Voltage-dependent block of tetrodotoxin-resistant (TTX-R) sodium currents.
  • Preferential selectivity versus NaV1.2 and NaV1.5 channels.
  • High purity suitable for research applications (≥97% HPLC).
Catalog No. 50-002-14989 Supplier Medchemexpress LLC Supplier No. HY10008010MG
A-887826 is a small-molecule, voltage-dependent NaV1.8 sodium channel blocker originally described by Abbott Laboratories. It is orally bioavailable, exhibits potent in vitro inhibition of NaV1.8 (IC50 ≈ 11 nM), and reduces neuropathic tactile allodynia in rodent models.
  • Potent NaV1.8 inhibition (IC50 ≈ 11 nM).
  • Orally bioavailable with demonstrated in vivo efficacy in neuropathic pain models.
  • Voltage-dependent block of tetrodotoxin-resistant (TTX-R) sodium currents.
  • Preferential selectivity versus NaV1.2 and NaV1.5 channels.
  • High purity suitable for research applications (≥97% HPLC).