Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5-(4-butylcyclohexyl)-3-pyridin-4-yl-1,2,4-oxadiazole hydrochloride | 388575-52-8 | MFCD01829706 | 100.0% | 285.38 g·mol⁻¹ | C17H23N3O | 50 MG
SDP

Catalog No. 5000242683
Encompass_Preferred

PSN 375963 is a small-molecule agonist of the G protein-coupled receptor GPR119 used in laboratory research to probe receptor function and signaling. It exhibits micromolar potency in human and mouse assays and is supplied as a hydrochloride salt with analytical data and storage recommendations for research use.

  • Potent GPR119 agonist with EC50 8.4 μM (human) and 7.9 μM (mouse).
  • High analytical purity reported at 99.96%.
  • Molecular formula C17H23N3O and molecular weight 285.38 g·mol⁻¹.
  • Available as hydrochloride salt and in solid pack sizes including 50 mg.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months).

Catalog No. 50-002-42683 Supplier Medchemexpress LLC Supplier No. HY10825850MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PSN 375963 is a small-molecule agonist of the G protein-coupled receptor GPR119 used in laboratory research to probe receptor function and signaling. It exhibits micromolar potency in human and mouse assays and is supplied as a hydrochloride salt with analytical data and storage recommendations for research use.

  • Potent GPR119 agonist with EC50 8.4 μM (human) and 7.9 μM (mouse).
  • High analytical purity reported at 99.96%.
  • Molecular formula C17H23N3O and molecular weight 285.38 g·mol⁻¹.
  • Available as hydrochloride salt and in solid pack sizes including 50 mg.
  • Recommended storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.