Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(4-Chlorobutyl)hydantoin 97.0+%, TCI America™
SDP

Catalog No. C09025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C09025G 5 g
C090225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C09025G Supplier TCI America Supplier No. C09025G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 40126-55-4
Molecular Formula C7H11ClN2O2
Molecular Weight (g/mol) 190.627
MDL Number MFCD00059722
InChI Key ZKHLVOULDIXSCC-UHFFFAOYSA-N
PubChem CID 229807
IUPAC Name 5-(4-chlorobutyl)imidazolidine-2,4-dione
SMILES C(CCCl)CC1C(=O)NC(=O)N1

Specifications

Melting Point 131°C
Color White-Yellow
Quantity 5 g
Formula Weight 190.63
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-(4-Chlorobutyl)hydantoin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.