Learn More
eMolecules​ 5-(4-Chlorophenyl)-1,3,4-oxadiazol-2-amine | 33621-61-3 | 5G | Purity: 95%
Supplier:  eMolecules​ 303361148
Encompass_Preferred
Combi-Blocks | 5-(4-Chlorophenyl)-1,3,4-oxadiazol-2-amine | 5G | 33621-61-3 | MFCD00461380
If you are unable to find the chemical you are looking for, make sure you are logged into your fishersci.com account and click on the following link:
eMolecules Building Block Tool
Catalog No. 50-159-8046