Learn More
Medchemexpress LLC 5-(4-Hydroxyphenyl)-3H-1,2-dithiole-3-thione | 18274-81-2 | 98.55% | 226.35 | 50 G
SDP
Catalog No. 5000344962
Encompass_Preferred
ADT-OH is a hydrogen sulfide-releasing donor that induces apoptosis and upregulates FADD. It inhibits FAK/Paxillin and shows potential for research in cancer diseases.
  • Induces apoptosis.
  • Upregulates FADD.
  • Inhibits FAK/Paxillin.
  • Significantly inhibits metastasis of TNBC cells.
  • Enhances antitumor activity of celecoxib in human colorectal cancer cells.
  • Promotes self-renewal and antiapoptosis ability of cultured neural precursor cells.
  • Exhibits anti-metastatic activity on malignant melanoma cells.
  • Inhibits human HDAC in HeLa cells.
  • Shows potent anti-metastatic activity in xenograft models.
  • Suppresses tumor metastasis with limited effect on primary tumor growth.
Catalog No. 50-003-44962 Supplier Medchemexpress LLC Supplier No. HY10958250MG
ADT-OH is a hydrogen sulfide-releasing donor that induces apoptosis and upregulates FADD. It inhibits FAK/Paxillin and shows potential for research in cancer diseases.
  • Induces apoptosis.
  • Upregulates FADD.
  • Inhibits FAK/Paxillin.
  • Significantly inhibits metastasis of TNBC cells.
  • Enhances antitumor activity of celecoxib in human colorectal cancer cells.
  • Promotes self-renewal and antiapoptosis ability of cultured neural precursor cells.
  • Exhibits anti-metastatic activity on malignant melanoma cells.
  • Inhibits human HDAC in HeLa cells.
  • Shows potent anti-metastatic activity in xenograft models.
  • Suppresses tumor metastasis with limited effect on primary tumor growth.