Learn More
Medchemexpress LLC 5-(4-Hydroxyphenyl)-3H-1,2-dithiole-3-thione | 18274-81-2 | 98.6% | 226.35 | 10 MG
SDP
Catalog No. 5000344440
Encompass_Preferred
ADT-OH is a hydrogen sulfide-releasing donor that induces apoptosis and upregulates FADD. It inhibits FAK/Paxillin and has potential for cancer disease research.
  • Functions as a hydrogen sulfide-releasing donor.
  • Induces programmed cell death.
  • Upregulates FADD.
  • Inhibits FAK/Paxillin.
  • Demonstrates potential for cancer disease research.
  • Significantly inhibits metastasis in various models.
  • Synergistically enhances the activity of celecoxib in human colorectal cancer cells.
  • Promotes self-renewal and antiapoptosis ability of cultured neural precursor cells.
  • Exhibits anti-metastatic activity on malignant melanoma cells through the inhibition of signaling pathways.
Catalog No. 50-003-44440 Supplier Medchemexpress LLC Supplier No. HY109582100MG
ADT-OH is a hydrogen sulfide-releasing donor that induces apoptosis and upregulates FADD. It inhibits FAK/Paxillin and has potential for cancer disease research.
  • Functions as a hydrogen sulfide-releasing donor.
  • Induces programmed cell death.
  • Upregulates FADD.
  • Inhibits FAK/Paxillin.
  • Demonstrates potential for cancer disease research.
  • Significantly inhibits metastasis in various models.
  • Synergistically enhances the activity of celecoxib in human colorectal cancer cells.
  • Promotes self-renewal and antiapoptosis ability of cultured neural precursor cells.
  • Exhibits anti-metastatic activity on malignant melanoma cells through the inhibition of signaling pathways.