Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(4-Methoxycarbonylphenyl)-10,15,20-triphenylporphyrin 90.0+%, TCI America™
SDP

Catalog No. M13381G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M13381G 1 g
M1338100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M13381G Supplier TCI America Supplier No. M13381G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 119730-06-2
Molecular Formula C46H32N4O2
Molecular Weight (g/mol) 672.788
MDL Number MFCD02093480
InChI Key JYTWPMXQCKOXJN-UHFFFAOYSA-N
Synonym 5-(4-Carbomethoxyphenyl)-10,15,20-triphenylporphyrin, 5-(4-Methoxycarbonylphenyl)-10,15,20-triphenyl-21H,23H-porphine
PubChem CID 11181451
IUPAC Name methyl 4-(10,15,20-triphenyl-23,24-dihydroporphyrin-5-yl)benzoate
SMILES COC(=O)C1=CC=C(C=C1)C2=C3C=CC(=N3)C(=C4C=CC(=C(C5=CC=C(N5)C(=C6C=CC2=N6)C7=CC=CC=C7)C8=CC=CC=C8)N4)C9=CC=CC=C9

Specifications

Color Blue-Purple
Quantity 1 g
Formula Weight 672.79
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-(4-Methoxycarbonylphenyl)-10,15,20-triphenylporphyrin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.