Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(4-Methoxyphenyl)-1H-tetrazole, 98%

Catalog No. AAL1721003
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1721003 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1721003 Supplier Thermo Scientific Chemicals Supplier No. L1721003

missing translation for 'validMainframeMsg'

CAS: 6926-51-8 | C8H8N4O | 176.179 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6926-51-8
Molecular Formula C8H8N4O
Molecular Weight (g/mol) 176.179
MDL Number MFCD00991875
InChI Key KZGGHPBTKGLWQL-UHFFFAOYSA-N
Synonym 5-4-methoxyphenyl-2h-tetrazole, 5-4-methoxy-phenyl-2h-tetrazole, 5-4-methoxyphenyl-1h-tetrazole, 2h-tetrazole,5-4-methoxyphenyl, 5-4-methoxyphenyl-2h-tetraazole, 5-4-methoxy-phenyl-1h-tetrazole, 5-4-methoxyphenyl-1h-1,2,3,4-tetrazole, 5-4-methoxyphenyl-2h-1,2,3,4-tetrazole, 1-1h-1,2,3,4-tetraazol-5-yl-4-methoxybenzene, 4-methoxyphenyl tetrazole
PubChem CID 307902
IUPAC Name 5-(4-methoxyphenyl)-2H-tetrazole
SMILES COC1=CC=C(C=C1)C2=NNN=N2

Specifications

Melting Point 235°C to 238°C
Quantity 1 g
Beilstein 152619
Formula Weight 176.18
Percent Purity 98%
Chemical Name or Material 5-(4-Methoxyphenyl)-1H-tetrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.