Learn More
Medchemexpress LLC 5-(4-nitrophenyl)-2-phenyl-4H-pyrazol-3-one | 34320-83-7 | 99.7% | 281.3 | C15H11N3O3 | 100 MG
SDP
Catalog No. 5000240255
Encompass_Preferred
TCS PrP Inhibitor 13 is a research small molecule that inhibits cellular prion protein (PrPC) and reduces accumulation of protease-resistant prion protein (PrP-res). It is employed in cellular studies to evaluate antiprion activity and related effects on cell viability and apoptosis.
  • Potent inhibitor with an IC50 of about 3 nM in ScN2a and F3 cells.
  • Reported to induce apoptosis in Schwannoma cells.
  • High purity (99.72%) suitable for research applications.
  • Supplied as a solid powder and as a DMSO solution (10 mM).
  • Stable when stored as a powder at -20 °C (up to 3 years) or 4 °C (up to 2 years).
  • In-solution storage: -80 °C (up to 6 months) or -20 °C (up to 1 month).
Catalog No. 50-002-40255 Supplier Medchemexpress LLC Supplier No. HY107662100MG
TCS PrP Inhibitor 13 is a research small molecule that inhibits cellular prion protein (PrPC) and reduces accumulation of protease-resistant prion protein (PrP-res). It is employed in cellular studies to evaluate antiprion activity and related effects on cell viability and apoptosis.
  • Potent inhibitor with an IC50 of about 3 nM in ScN2a and F3 cells.
  • Reported to induce apoptosis in Schwannoma cells.
  • High purity (99.72%) suitable for research applications.
  • Supplied as a solid powder and as a DMSO solution (10 mM).
  • Stable when stored as a powder at -20 °C (up to 3 years) or 4 °C (up to 2 years).
  • In-solution storage: -80 °C (up to 6 months) or -20 °C (up to 1 month).