Learn More
Chemglass Life Sciences 3.50" ID x 5.45" OD x 1.20" Height White PTFE Support Ring
SDP
Catalog No. 50872570
Encompass_Preferred
3.50" ID x 5.45" OD x 1.20" Height White PTFE Support Ring. PTFE support ring used to support round bottom flasks or cylindrical reaction vessels.PLEASE NOTE: PTFE SUPPORT RINGS CAN BE CUT. PLEASE CALL FOR PRICING
Catalog No. 50-872-570 Supplier Chemglass Life Sciences Supplier No. CG305505
3.50" ID x 5.45" OD x 1.20" Height White PTFE Support Ring. PTFE support ring used to support round bottom flasks or cylindrical reaction vessels.PLEASE NOTE: PTFE SUPPORT RINGS CAN BE CUT. PLEASE CALL FOR PRICING