Learn More
Medchemexpress LLC 5,5'-(((4-chloro-1,2-phenylene)bis(azanediyl))bis(methylene))bis(2-methylphenol) | 2863686-81-9 | 99.6% | C22H23ClN2O2 | 10MG
SDP
Catalog No. 5000258864
Encompass_Preferred
DRB18 is a small-molecule pan-class inhibitor of glucose transporters (GLUTs) used in vitro to modulate cellular glucose uptake and metabolism. It alters glucose-related metabolites, can induce G1/S phase arrest, increase oxidative stress, and promote necrotic cell death in treated cell lines. Supplied as a white to yellow solid for research use.
  • Pan-class GLUT inhibition for metabolic and cancer research.
  • Alters glucose-related metabolites and energy pathways.
  • Reported to induce cell cycle arrest and oxidative stress.
  • High purity for reproducible experimental results.
  • Solid form suitable for long-term storage under recommended conditions.
Catalog No. 50-002-58864 Supplier Medchemexpress LLC Supplier No. HY14596310MG
DRB18 is a small-molecule pan-class inhibitor of glucose transporters (GLUTs) used in vitro to modulate cellular glucose uptake and metabolism. It alters glucose-related metabolites, can induce G1/S phase arrest, increase oxidative stress, and promote necrotic cell death in treated cell lines. Supplied as a white to yellow solid for research use.
  • Pan-class GLUT inhibition for metabolic and cancer research.
  • Alters glucose-related metabolites and energy pathways.
  • Reported to induce cell cycle arrest and oxidative stress.
  • High purity for reproducible experimental results.
  • Solid form suitable for long-term storage under recommended conditions.