Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5,5-Trifluoro-DL-leucine, 94%, Thermo Scientific™

Catalog No. AAL12890ME
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL12890ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL12890ME Supplier Thermo Scientific Chemicals Supplier No. L12890ME

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5,5,5-Trifluoro-DL-leucine is used as an intermediate in chemical research.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Store at -80°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2792-72-5
Molecular Formula C6H10F3NO2
Molecular Weight (g/mol) 185.146
MDL Number MFCD00039528
InChI Key XFGVJLGVINCWDP-UHFFFAOYSA-N
Synonym 5,5,5-trifluoro-dl-leucine, 5,5,5-trifluoroleucine, 5',5',5'-trifluoroleucine, dl-5',5',5'-trifluoroleucine, leucine,5,5,5-trifluoro, leucine, 5,5,5-trifluoro, 2-azanyl-5,5,5-tris fluoranyl-4-methyl-pentanoic acid, acmc-1cocp, norvaline, 5,5,5-trifluoro-4-methyl-, dl, dl-leucine, 5,5,5-trifluoro
PubChem CID 259832
ChEBI CHEBI:78014
IUPAC Name 2-amino-5,5,5-trifluoro-4-methylpentanoic acid
SMILES CC(CC(C(=O)O)N)C(F)(F)F

Specifications

Melting Point 273°C to 275°C
Beilstein 4664821
Quantity 500 mg
Formula Weight 185.15
Percent Purity 94%
Chemical Name or Material 5,5,5-Trifluoro-DL-leucine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.