Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5'-Bis(trifluoromethyl)-2,2'-bipyridyl 98.0+%, TCI America™
SDP

Catalog No. B5449200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5449200MG 200 mg
B54491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5449200MG Supplier TCI America Supplier No. B5449200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 142946-80-3
Molecular Formula C12H6F6N2
Molecular Weight (g/mol) 292.18
MDL Number MFCD24449717
InChI Key ZHMXQYAUGQASQM-UHFFFAOYSA-N
Synonym 5,5′-Bis(trifluoromethyl)-2,2′-bipyridine
PubChem CID 589903
IUPAC Name 5,5'-bis(trifluoromethyl)-2,2'-bipyridine
SMILES FC(F)(F)C1=CN=C(C=C1)C1=NC=C(C=C1)C(F)(F)F

Specifications

Melting Point 131°C
Color White-Yellow
Quantity 200 mg
Formula Weight 292.18
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5,5′-Bis(trifluoromethyl)-2,2′-bipyridyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.