Learn More
Medchemexpress LLC 5-(5-bromo-2-((2-bromobenzyl)oxy)benzylidene)-2-thioxothiazolidin-4-one | 893449-38-2 | MFCD11045303 | 99.1% | 485.21 | C17H11Br2NO2S2 | 5 MG
SDP
Catalog No. 5000263457
Encompass_Preferred
PRL-3 Inhibitor I is a small-molecule research reagent that inhibits the phosphatase PRL-3 (PTP4A3) and is used to probe PRL-3-dependent signaling and cellular invasion. The compound is provided as a high-purity solid; DMSO is the recommended solvent for preparing assay stock solutions.
  • Potent PRL-3 inhibition with IC50 0.9 μM.
  • High purity suitable for biochemical and cell-based assays.
  • Molecular formula C17H11Br2NO2S2; molecular weight 485.21.
  • Available in small milligram quantities and as DMSO stock solutions for convenient assay setup.
  • Supplied for research use only; not for human or clinical use.
Catalog No. 50-002-63457 Supplier Medchemexpress LLC Supplier No. HY1124765MG
PRL-3 Inhibitor I is a small-molecule research reagent that inhibits the phosphatase PRL-3 (PTP4A3) and is used to probe PRL-3-dependent signaling and cellular invasion. The compound is provided as a high-purity solid; DMSO is the recommended solvent for preparing assay stock solutions.
  • Potent PRL-3 inhibition with IC50 0.9 μM.
  • High purity suitable for biochemical and cell-based assays.
  • Molecular formula C17H11Br2NO2S2; molecular weight 485.21.
  • Available in small milligram quantities and as DMSO stock solutions for convenient assay setup.
  • Supplied for research use only; not for human or clinical use.