Learn More
Medchemexpress LLC 5-((5-chloro-2-methoxyphenyl)sulfonamido)nicotinamide | 1451272-71-1 | 98.9% | 341.77 | C13H12ClN3O4S | 100MG
SDP
Catalog No. 5000236291
Encompass_Preferred
SBI-425 is a potent, selective, orally bioavailable tissue-nonspecific alkaline phosphatase (TNAP) inhibitor supplied for research use. It is provided as a high-purity solid with defined molecular properties and is offered in multiple package sizes for laboratory and preclinical studies. Not for human use.
  • Potent and selective TNAP inhibitor.
  • Orally bioavailable suitable for in vivo research.
  • High purity (98.9%).
  • Available in multiple package sizes including 100 mg.
  • Solid powder with molecular weight 341.77 and formula C13H12ClN3O4S.
  • For research use only; not for human therapeutic use.
Catalog No. 50-002-36291 Supplier Medchemexpress LLC Supplier No. HY124756100MG
SBI-425 is a potent, selective, orally bioavailable tissue-nonspecific alkaline phosphatase (TNAP) inhibitor supplied for research use. It is provided as a high-purity solid with defined molecular properties and is offered in multiple package sizes for laboratory and preclinical studies. Not for human use.
  • Potent and selective TNAP inhibitor.
  • Orally bioavailable suitable for in vivo research.
  • High purity (98.9%).
  • Available in multiple package sizes including 100 mg.
  • Solid powder with molecular weight 341.77 and formula C13H12ClN3O4S.
  • For research use only; not for human therapeutic use.