Learn More
Medchemexpress LLC 5-((5-chloro-2-methoxyphenyl)sulfonamido)nicotinamide | 1451272-71-1 | 98.9% | 341.77 g/mol | C13H12ClN3O4S | 10MG
SDP
Catalog No. 5000235219
Encompass_Preferred
SBI-425 is a small-molecule tissue-nonspecific alkaline phosphatase (TNAP) inhibitor intended for research use. It is reported as a potent, selective, and orally bioavailable compound developed for use in mechanistic studies and in vivo models.
  • Inhibits tissue-nonspecific alkaline phosphatase (TNAP).
  • High stated purity (~98.9%).
  • Molecular weight 341.77 g/mol and formula C13H12ClN3O4S.
  • Available in multiple solid sizes and as DMSO solutions for ready use.
  • Suitable for in vitro and in vivo research applications.
Catalog No. 50-002-35219 Supplier Medchemexpress LLC Supplier No. HY12475610MG
SBI-425 is a small-molecule tissue-nonspecific alkaline phosphatase (TNAP) inhibitor intended for research use. It is reported as a potent, selective, and orally bioavailable compound developed for use in mechanistic studies and in vivo models.
  • Inhibits tissue-nonspecific alkaline phosphatase (TNAP).
  • High stated purity (~98.9%).
  • Molecular weight 341.77 g/mol and formula C13H12ClN3O4S.
  • Available in multiple solid sizes and as DMSO solutions for ready use.
  • Suitable for in vitro and in vivo research applications.