Learn More
Medchemexpress LLC 5-[(5-chloro-2-methoxyphenyl)sulfonylamino]pyridine-3-carboxamide | 1451272-71-1 | 98.9% | 341.77 g·mol⁻¹ | C13H12ClN3O4S | 50MG
SDP
Catalog No. 5000235842
Encompass_Preferred
SBI-425 is a small-molecule, orally bioavailable inhibitor of tissue-nonspecific alkaline phosphatase (TNAP) used in preclinical research to modulate vascular TNAP activity and study cardiovascular and calcification pathways. It is supplied as a solid with defined molecular properties and high purity for use in in vitro and in vivo experiments.
  • Orally bioavailable TNAP inhibitor with reported IC50 ≈ 16 nM.
  • High purity suitable for research use (≈98.9%).
  • Molecular formula C13H12ClN3O4S and molecular weight 341.77 g·mol⁻¹.
  • Supplied as a solid for handling in laboratory workflows.
  • Available in multiple small package sizes for preclinical studies.
  • Intended for research use only; not for human or diagnostic use.
Catalog No. 50-002-35842 Supplier Medchemexpress LLC Supplier No. HY12475650MG
SBI-425 is a small-molecule, orally bioavailable inhibitor of tissue-nonspecific alkaline phosphatase (TNAP) used in preclinical research to modulate vascular TNAP activity and study cardiovascular and calcification pathways. It is supplied as a solid with defined molecular properties and high purity for use in in vitro and in vivo experiments.
  • Orally bioavailable TNAP inhibitor with reported IC50 ≈ 16 nM.
  • High purity suitable for research use (≈98.9%).
  • Molecular formula C13H12ClN3O4S and molecular weight 341.77 g·mol⁻¹.
  • Supplied as a solid for handling in laboratory workflows.
  • Available in multiple small package sizes for preclinical studies.
  • Intended for research use only; not for human or diagnostic use.