Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5‴-Di-n-octyl-2,2':5',2″:5″,2‴-quaterthi 98+%, TCI America™
SDP

Catalog No. D4877100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4877100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4877100MG Supplier TCI America Supplier No. D4877100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 882659-01-0
Molecular Formula C32H42S4
Molecular Weight (g/mol) 554.928
InChI Key XUXLXUSOBDWJBD-UHFFFAOYSA-N
PubChem CID 58892812
IUPAC Name 2-octyl-5-[5-[5-(5-octylthiophen-2-yl)thiophen-2-yl]thiophen-2-yl]thiophene
SMILES CCCCCCCCC1=CC=C(S1)C2=CC=C(S2)C3=CC=C(S3)C4=CC=C(S4)CCCCCCCC

Specifications

Melting Point 176°C
Quantity 100 mg
Formula Weight 554.93
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5,5′′′-Di-n-octyl-2,2′:5′,2′′:5′′,2′′
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.