Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5'-Dibromo-BAPTA, 95%, Thermo Scientific™

Catalog No. AAA1138803
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1138803 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1138803 Supplier Thermo Scientific Chemicals Supplier No. A1138803

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 73630-11-2
Molecular Formula C22H22Br2N2O10
Quantity 1g
MDL Number MFCD00036169
Synonym dibromo-bapta, 5,5'-dibromo-bapta, 5,5'-dibromo-1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, br2-bapta, 1,2-bis 2-bis carboxymethyl amino-5-bromophenoxy ethane, 1,2-bis 2-bis 2-amino-5-bromophenoxy ethane-n,n,n',n'-tetraacetic acid, 1,2-bis 2-amino-5-bromophenoxy ethane-n,n,n',n'-tetraacetic acid, 2,2',2,2'-ethane-1,2-diylbis oxy 4-bromo-2,1-phenylene nitrilo tetraacetic acid, n,n'-1,2-ethanediylbis oxy 4-bromo-2,1-phenylene bis n-carboxymethyl glycine, dibromobapta
InChI Key KCTRRYZOCJDOTC-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1Br)OCCOC2=C(C=CC(=C2)Br)N(CC(=O)O)CC(=O)O)N(CC(=O)O)CC(=O)O
IUPAC Name 2-[2-[2-[2-[bis(carboxymethyl)amino]-5-bromophenoxy]ethoxy]-4-bromo-N-(carboxymethyl)anilino]acetic acid
Molecular Weight (g/mol) 634.23
PubChem CID 3081001
ChEBI CHEBI:60890
Formula Weight 634.23
Percent Purity 95%
Chemical Name or Material 5,5'-Dibromo-BAPTA
Show More Show Less
Recommended Storage 2°C to 8°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.