Learn More
Medchemexpress LLC 5,5'-Dicarboxy-2,2'-bipyridine | 1802-30-8 | 244.20 | 1 G
SDP
Catalog No. 5000445465
Encompass_Preferred
5,5'-Dicarboxy-2,2'-bipyridine is a niacin derivative with potential applications in the research of conditions such as hypercholesterolemia, hyperlipidemia, and cardiovascular disease. This chemical compound is provided as an off-white to gray solid and is characterized by its molecular formula C₁₂H₈N₂O₄ and a molecular weight of 244.20.
  • High purity of 99.76% (HPLC)
  • Promising for research of hypercholesterolemia
  • Promising for research of hyperlipidemia
  • Promising for research of cardiovascular disease
  • Consistent with structure by ¹H NMR spectroscopy
  • Soluble in DMSO: 2 mg/mL
Catalog No. 50-004-45465 Supplier Medchemexpress LLC Supplier No. HYW0045801G
5,5'-Dicarboxy-2,2'-bipyridine is a niacin derivative with potential applications in the research of conditions such as hypercholesterolemia, hyperlipidemia, and cardiovascular disease. This chemical compound is provided as an off-white to gray solid and is characterized by its molecular formula C₁₂H₈N₂O₄ and a molecular weight of 244.20.
  • High purity of 99.76% (HPLC)
  • Promising for research of hypercholesterolemia
  • Promising for research of hyperlipidemia
  • Promising for research of cardiovascular disease
  • Consistent with structure by ¹H NMR spectroscopy
  • Soluble in DMSO: 2 mg/mL