Learn More
Medchemexpress LLC 5,5'-Dicarboxy-2,2'-bipyridine | 1802-30-8 | 244.20 | 10 MG
SDP
Catalog No. 5000445467
Encompass_Preferred
5,5'-Dicarboxy-2,2'-bipyridine is a niacin derivative with potential applications in hypercholesterolemia, hyperlipidemia, and cardiovascular disease research.
  • Promising for research of hypercholesterolemia
  • Promising for research of hyperlipidemia
  • Promising for research of cardiovascular disease
Catalog No. 50-004-45467 Supplier Medchemexpress LLC Supplier No. HYW00458010G
5,5'-Dicarboxy-2,2'-bipyridine is a niacin derivative with potential applications in hypercholesterolemia, hyperlipidemia, and cardiovascular disease research.
  • Promising for research of hypercholesterolemia
  • Promising for research of hyperlipidemia
  • Promising for research of cardiovascular disease