Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,5'-Dicarboxy-2,2'-bipyridine | 1802-30-8 | 244.20 | 5 G
SDP

Catalog No. 5000445395
Encompass_Preferred

5,5'-Dicarboxy-2,2'-bipyridine is an off-white to gray solid and a niacin derivative that shows promise in research related to hypercholesterolemia, hyperlipidemia, and cardiovascular disease.

  • High purity (99.8% by HPLC)
  • Stable under recommended storage conditions
  • Recommended storage at 4°C, protected from light and under nitrogen
  • Soluble in DMSO (2 mg/mL)
  • Suitable for research on hypercholesterolemia, hyperlipidemia, and cardiovascular disease

Catalog No. 50-004-45395 Supplier Medchemexpress LLC Supplier No. HYW0045805G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5,5'-Dicarboxy-2,2'-bipyridine is an off-white to gray solid and a niacin derivative that shows promise in research related to hypercholesterolemia, hyperlipidemia, and cardiovascular disease.

  • High purity (99.8% by HPLC)
  • Stable under recommended storage conditions
  • Recommended storage at 4°C, protected from light and under nitrogen
  • Soluble in DMSO (2 mg/mL)
  • Suitable for research on hypercholesterolemia, hyperlipidemia, and cardiovascular disease

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.