Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,5'-Dicarboxy-2,2'-bipyridine | 1802-30-8 | 244.20 | 50 G
SDP

Catalog No. 5000448769
Encompass_Preferred

5,5'-Dicarboxy-2,2'-bipyridine is a niacin derivative that shows promise for research into hypercholesterolemia, hyperlipidemia, and cardiovascular disease.

  • Storage at 4°C, protected from light, and under nitrogen.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen.
  • Targeted for MOFs.
  • Appearance: Off-white to gray solid.

Catalog No. 50-004-48769 Supplier Medchemexpress LLC Supplier No. HYW00458050G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5,5'-Dicarboxy-2,2'-bipyridine is a niacin derivative that shows promise for research into hypercholesterolemia, hyperlipidemia, and cardiovascular disease.

  • Storage at 4°C, protected from light, and under nitrogen.
  • In solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light and under nitrogen.
  • Targeted for MOFs.
  • Appearance: Off-white to gray solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.