Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-(+)-5,5'-Dichloro-2,2'-Bis(Diphenylphosphino)-6,6'-Dimethoxy-1,1'-Biphenyl

Catalog No. 96738250MGF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
96-738-250MGF 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 96-738-250MGF Supplier Sigma Aldrich Supplier No. 96738250MGF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Transition metal complexes are used as catalysts for asymmetric hydrogenation; Eur Pat Appl 749973, 1995 (20.06.1995); Eur Pat Appl 1160237 (18.05.2001).produced by Bayer

Specifications

CAS 59158-14-4
Melting Point 61°C to 65°C
Molecular Formula C25H55NO4S
Linear Formula [CH3(CH2)7]3N(OSO3H)CH3
MDL Number MFCD03095545
Quantity 5 g
Molecular Weight (g/mol) 465.77 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.