Learn More
AdipoGen 5,5'-Difluoro BAPTA-AM (50 mg)
SDP
Supplier:  AdipoGen CDXB0445M050
Encompass_Preferred
Chemical. CAS: 128255-42-5. Formula: C34H38F2N2O18. MW: 800.66. 5,5′-Difluoro-BAPTA-AM is a calcium chelator that may be used to modulate calcium levels and flow within cells to study calcium homeostasis and cell signaling. It is used to study cytosolic free Ca2+ by 19F-NMR. It has high selectivity for Ca2+, it is not particularly pH sensitive and is available protected as its acetoxymethyl ester. Determination with 5,5′-Difluoro-BAPTA-AM are not sensitive to cells which have a high natural background fluorescence.
Catalog No. 50-205-4321