Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5'-Diiodo-2,2'-dimethylbiphenyl 97.0+%, TCI America™
SDP

Catalog No. D54301G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D54301G 1 g
D54305G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D54301G Supplier TCI America Supplier No. D54301G
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1392203-19-8
Molecular Formula C14H12I2
Molecular Weight (g/mol) 434.06
MDL Number MFCD30742904
InChI Key VBRWVADZBWZCMH-UHFFFAOYSA-N
PubChem CID 126728034
IUPAC Name 5,5'-diiodo-2,2'-dimethyl-1,1'-biphenyl
SMILES CC1=C(C=C(I)C=C1)C1=C(C)C=CC(I)=C1

Specifications

Melting Point 117°C
Color White
Quantity 1 g
Formula Weight 434.06
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5,5′-Diiodo-2,2′-dimethylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.