Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5-Dimethyl-1,3-cycloHexanedione, For HPLC derivatization, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111012870 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-2870 250 g
11-101-2872 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-2870 Supplier MilliporeSigma Supelco Supplier No. 38490250G
Add to Cart
Add to Cart

5,5-dimethyl-1,3-cyclohexanedione (Dimedone) is basically a highly sensitive and very specific reagent used for the determination of aldehydes. It is highly advantageous as it is environment friendly, give high yields and is a simple workup procedure.

General Analytical
TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00001588
Synonym Dimedone; Methone

Specifications

Melting Point 146°C to 148°C
CAS 126-81-8
Linear Formula (CH3)2C6H6(=O)2
Quantity 250 g
Formula Weight 140.18
Percent Purity ≥99.0% (GC); ≥99.0%
Grade For HPLC derivatization
RTECSNumber GV0345000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.