Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5'-Dimethyl-2,2'-bipyridyl 98.0+%, TCI America™
SDP

Catalog No. B21381G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21381G 1 g
B21385G 5 g
B213825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B21381G Supplier TCI America Supplier No. B21381G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1762-34-1
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.242
MDL Number MFCD01740554
InChI Key PTRATZCAGVBFIQ-UHFFFAOYSA-N
Synonym 5,5'-dimethyl-2,2'-bipyridine, abametapir, 5,5'-dimethyl-2,2'-dipyridyl, 6,6'-bi-3-picoline, 5,5'-dimethyl-2,2'-bipyridyl, 6,6'-di-3-picolyl, 6,6'-di-3-picoline, unii-6uo390amfb, 2,2'-bipyridine, 5,5'-dimethyl, 5,5-dimethyl-2,2-dipyridyl
PubChem CID 15664
IUPAC Name 5-methyl-2-(5-methylpyridin-2-yl)pyridine
SMILES CC1=CN=C(C=C1)C2=NC=C(C=C2)C

Specifications

Melting Point 115°C
Quantity 1 g
Formula Weight 184.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5,5′-Dimethyl-2,2′-bipyridyl
RTECSNumber DW1766000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.