Learn More
Medchemexpress LLC 5,5-dimethylcyclohexane-1,3-dione | 126-81-8 | MFCD00001588 | 99.7% | 140.18 g/mol | C8H12O2 | 500 G
SDP
Catalog No. 5000300013
Encompass_Preferred
Dimedone (5,5-dimethylcyclohexane-1,3-dione) is a small-molecule biochemical reagent used in organic synthesis and life-science research. It is commonly used as a nucleophilic building block and as a selective probe for sulfenic acid modification in proteins; supplied as a high-purity solid with supporting analytical documentation.
  • High purity (99.7%).
  • Solid form, easy to weigh and store.
  • Selective reactivity with sulfenic acids for biochemical labeling.
  • Suitable for organic synthesis and biochemical assays.
  • Certificate of analysis and safety data sheet available.
Catalog No. 50-003-00013 Supplier Medchemexpress LLC Supplier No. HYW015740500G
Dimedone (5,5-dimethylcyclohexane-1,3-dione) is a small-molecule biochemical reagent used in organic synthesis and life-science research. It is commonly used as a nucleophilic building block and as a selective probe for sulfenic acid modification in proteins; supplied as a high-purity solid with supporting analytical documentation.
  • High purity (99.7%).
  • Solid form, easy to weigh and store.
  • Selective reactivity with sulfenic acids for biochemical labeling.
  • Suitable for organic synthesis and biochemical assays.
  • Certificate of analysis and safety data sheet available.