Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5,5-Diphenylhydantoin sodium salt, 99%

Catalog No. p-4258709
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC436100250 25 g Glass bottle
AC436101000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC436100250 Supplier Thermo Scientific Chemicals Supplier No. 436100250
Only null left
Add to Cart

CAS: 630-93-3 | C15H11N2NaO2 | 274.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-93-3
Molecular Formula C15H11N2NaO2
Molecular Weight (g/mol) 274.26
MDL Number MFCD00069674,MFCD00005264
InChI Key FJPYVLNWWICYDW-UHFFFAOYSA-M
Synonym Phenytoin sodium, Sodium 5, 5-diphenyl-2, 4-imidazolidinedione
PubChem CID 131674077
SMILES [Na+].O=C1NC(C(=O)[N-]1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 295°C to 298°C
Color White
CAS Min % 98.5
CAS Max % 100.0
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 99%
Merck Index 15,7433
Solubility Information Solubility in water: practically insoluble
IUPAC Name sodium 2,5-dioxo-4,4-diphenylimidazolidin-1-ide
Formula Weight 274.25
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material 5, 5-Diphenylhydantoin sodium salt
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.
Suspected of causing cancer.
Suspected of damaging fertility.
Suspected of damaging the unborn child.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: G
EINECSNumber 211-148-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.