Learn More
Chemglass Life Sciences 5.5in Ground Flat Flange, Tube ID: 130mm, Tube OD: 140mm
SDP
Catalog No. CG13505
Encompass_Preferred
Mating end of flange has a ground surface to provide for a liquid tight seal when greased and clamped with the CG-139 horseshoe clamp. 5.5in Ground Flat Flange, Tube ID: 130mm, Tube OD: 140mm.
Catalog No. CG13505 Supplier Chemglass Life Sciences Supplier No. CG13505
Mating end of flange has a ground surface to provide for a liquid tight seal when greased and clamped with the CG-139 horseshoe clamp. 5.5in Ground Flat Flange, Tube ID: 130mm, Tube OD: 140mm.