Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(5-Methyl-2-thienyl)thieno[2,3-d]pyrimidin-4-one, 97%, Thermo Scientific Chemicals

Catalog No. AAH5885603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5885603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5885603 Supplier Thermo Scientific Chemicals Supplier No. H5885603
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 851116-03-5 | Molecular Formula: C11H8N2OS2 | Molecular Weight: 248.318 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 851116-03-5
Molecular Formula C11H8N2OS2
Molecular Weight (g/mol) 248.318
MDL Number MFCD06671220
InChI Key SYSIILLIYRVOMM-UHFFFAOYSA-N
Synonym 5-5-methylthiophen-2-yl-3h-thieno 2,3-d pyrimidin-4-one, 5-5-methylthiophen-2-yl-3h,4h-thieno 2,3-d pyrimidin-4-one, 5-5-methyl-2-thienyl thieno 2,3-d pyrimidin-4-one
PubChem CID 4963645
IUPAC Name 5-(5-methylthiophen-2-yl)-3H-thieno[2,3-d]pyrimidin-4-one
SMILES CC1=CC=C(S1)C2=CSC3=C2C(=O)NC=N3

Specifications

Quantity 1 g
Formula Weight 248.31
Percent Purity 97%
Chemical Name or Material 5-(5-Methyl-2-thienyl)thieno[2,3-d]pyrimidin-4-one
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.