Learn More
Medchemexpress LLC 5-[5-phenyl-4-(pyridin-2-ylmethylamino)quinazolin-2-yl]pyridine-3-sulfonamide | 1272353-82-8 | 98.0% | 1 MG
SDP
Catalog No. 5000234829
Encompass_Preferred
BMS-919373 is a selective, potent IKur (Kv1.5) channel blocker used in cardiovascular research, including studies of atrial electrophysiology. It is provided as a research-grade powder with characterized purity and stability.
  • Selective IKur (Kv1.5) channel blocker with cardiovascular research applications.
  • Demonstrated in vitro activity (HEK293 hERG IC50 = 1.9 μM by patch clamp assay).
  • Chemical formula C25H20N6O2S and molecular weight 468.53 g/mol.
  • Available in small research pack sizes and stored frozen for long-term stability.
  • Reported supplier purity approximately 98%, suitable for laboratory research.
Catalog No. 50-002-34829 Supplier Medchemexpress LLC Supplier No. HY1206441MG
BMS-919373 is a selective, potent IKur (Kv1.5) channel blocker used in cardiovascular research, including studies of atrial electrophysiology. It is provided as a research-grade powder with characterized purity and stability.
  • Selective IKur (Kv1.5) channel blocker with cardiovascular research applications.
  • Demonstrated in vitro activity (HEK293 hERG IC50 = 1.9 μM by patch clamp assay).
  • Chemical formula C25H20N6O2S and molecular weight 468.53 g/mol.
  • Available in small research pack sizes and stored frozen for long-term stability.
  • Reported supplier purity approximately 98%, suitable for laboratory research.