Learn More
Scilogex LLC MicroPette Single Channel Variable Volume Pipettors, 5-50 uL
Catalog No. NC1747098
Encompass_Preferred
Lightweight and ergonomic design. Clear digital display. Integral tip ejector for access into narrow tubes. Easy to calibrate and maintain with tool supplied. Autoclavable tip-cone. Used with 'universal' pipette tips.
Catalog No. NC1747098 Supplier Scilogex LLC Supplier No. 712111069999
Lightweight and ergonomic design. Clear digital display. Integral tip ejector for access into narrow tubes. Easy to calibrate and maintain with tool supplied. Autoclavable tip-cone. Used with 'universal' pipette tips.