Learn More
Medchemexpress LLC 5-[6-[[(3R)-1-azabicyclo[2.2.2]octan-3-yl]oxy]pyridazin-3-yl]-1H-indole | 855291-54-2 | 99.7% | 320.39 g/mol | C19H20N4O | 10 MG
SDP
Catalog No. 5000242979
Encompass_Preferred
ABT-107 is a selective α7 neuronal nicotinic acetylcholine receptor agonist used in preclinical research to probe α7-mediated signaling and neuroprotection. It has shown protection against nigrostriatal damage in rodent models and is supplied for both in vitro and in vivo experimental applications.
  • Selective α7 neuronal nicotinic receptor agonist suitable for receptor pharmacology studies.
  • Demonstrated neuroprotective effects in rodent nigrostriatal lesion models.
  • White to off-white solid for use in analytical and biological assays.
  • High purity (approximately 99.7%) for reproducible experimental results.
  • Soluble in DMSO at 100 mg/mL; recommended in vivo formulation protocols available.
  • Stable when stored as a powder at -20°C; follow solvent storage guidance for solutions.
  • Available as solid and as pre-made DMSO solutions for convenient dosing.
Catalog No. 50-002-42979 Supplier Medchemexpress LLC Supplier No. HY10803810MG
ABT-107 is a selective α7 neuronal nicotinic acetylcholine receptor agonist used in preclinical research to probe α7-mediated signaling and neuroprotection. It has shown protection against nigrostriatal damage in rodent models and is supplied for both in vitro and in vivo experimental applications.
  • Selective α7 neuronal nicotinic receptor agonist suitable for receptor pharmacology studies.
  • Demonstrated neuroprotective effects in rodent nigrostriatal lesion models.
  • White to off-white solid for use in analytical and biological assays.
  • High purity (approximately 99.7%) for reproducible experimental results.
  • Soluble in DMSO at 100 mg/mL; recommended in vivo formulation protocols available.
  • Stable when stored as a powder at -20°C; follow solvent storage guidance for solutions.
  • Available as solid and as pre-made DMSO solutions for convenient dosing.