Learn More
Medchemexpress LLC 5-[6-[[(3R)-1-azabicyclo[2.2.2]octan-3-yl]oxy]pyridazin-3-yl]-1H-indole | 855291-54-2 | 99.7% | 320.39 g/mol | C19H20N4O | 50 MG
SDP
Catalog No. 5000243023
Encompass_Preferred
ABT-107 is a selective α7 neuronal nicotinic receptor agonist for preclinical pharmacology and neuroscience research. It has demonstrated neuroprotective effects in rodent models of nigrostriatal injury and is supplied as a high-purity research reagent with recommended formulations for in vitro and in vivo studies.
  • Selective α7 neuronal nicotinic receptor agonist.
  • Demonstrated neuroprotection in 6-hydroxydopamine rodent model.
  • High purity (99.73%).
  • Molecular formula C19H20N4O; molecular weight 320.39 g/mol.
  • Available as solid and DMSO solution for experimental use.
  • Recommended solvents for in vitro and in vivo use, including DMSO and PEG-based formulations.
Catalog No. 50-002-43023 Supplier Medchemexpress LLC Supplier No. HY10803850MG
ABT-107 is a selective α7 neuronal nicotinic receptor agonist for preclinical pharmacology and neuroscience research. It has demonstrated neuroprotective effects in rodent models of nigrostriatal injury and is supplied as a high-purity research reagent with recommended formulations for in vitro and in vivo studies.
  • Selective α7 neuronal nicotinic receptor agonist.
  • Demonstrated neuroprotection in 6-hydroxydopamine rodent model.
  • High purity (99.73%).
  • Molecular formula C19H20N4O; molecular weight 320.39 g/mol.
  • Available as solid and DMSO solution for experimental use.
  • Recommended solvents for in vitro and in vivo use, including DMSO and PEG-based formulations.