Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 5,6,7,8-tetrahydroquinolin-2(1H)-one | 54802-19-6 | MFCD00456287 | >98.0% | 149.19 g/mol | C9H11NO | 5G
SDP

Catalog No. 5000237572
Encompass_Preferred

5,6,7,8-Tetrahydro-2-quinolone (NSC 99006) is a small-molecule synthetic intermediate used in pharmaceutical and medicinal chemistry research. It is provided for research use and serves as a building block for synthesis and method development.

  • Synthetic intermediate for pharmaceutical synthesis.
  • CAS number 54802-19-6 for unambiguous identification.
  • Molecular formula C9H11NO.
  • Molecular weight 149.19 g/mol.
  • Available in multiple pack sizes, including 5 g.
  • Supplied for research use only; not for human or veterinary use.

Catalog No. 50-002-37572 Supplier Medchemexpress LLC Supplier No. HY1230295G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5,6,7,8-Tetrahydro-2-quinolone (NSC 99006) is a small-molecule synthetic intermediate used in pharmaceutical and medicinal chemistry research. It is provided for research use and serves as a building block for synthesis and method development.

  • Synthetic intermediate for pharmaceutical synthesis.
  • CAS number 54802-19-6 for unambiguous identification.
  • Molecular formula C9H11NO.
  • Molecular weight 149.19 g/mol.
  • Available in multiple pack sizes, including 5 g.
  • Supplied for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.