Learn More
Cayman Chemical 5,6,7-Trimethoxyflavone, Purity: ≥95%, CAS: 973-67-1, 100 mg
SDP
Catalog No. 502943713
Encompass_Preferred
A flavonoid with diverse biological activities; reduces the viability of HepG2 and Hep3B carcinoma cells at 50 µM; decreases HSV-1 and poliovirus replication in HSV-1- or poliovirus-infected Vero cells, as well as decreases CMV replication in CMV-infected MRC-5 cells (IC50s = 3.2, 32, and 8 mg/L, respectively); inhibits LPS-induced NO, PGE2, Tnf-α, Il-1β, and Il-6 production in RAW 264.7 macrophages at 20 or 40 µM; increases survival in a mouse model of LPS-induced sepsis at 40 mg/kg
Catalog No. 50-294-3713 Supplier Cayman Chemical Supplier No. 40289100 MG
A flavonoid with diverse biological activities; reduces the viability of HepG2 and Hep3B carcinoma cells at 50 µM; decreases HSV-1 and poliovirus replication in HSV-1- or poliovirus-infected Vero cells, as well as decreases CMV replication in CMV-infected MRC-5 cells (IC50s = 3.2, 32, and 8 mg/L, respectively); inhibits LPS-induced NO, PGE2, Tnf-α, Il-1β, and Il-6 production in RAW 264.7 macrophages at 20 or 40 µM; increases survival in a mouse model of LPS-induced sepsis at 40 mg/kg