Learn More
TARGETMOL CHEMICALS INC 5 6 7-TRIMETHOXYFLAVONE 25MG
SDP
Catalog No. 502878219
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 5,6,7-Trimethoxyflavone, a natural product extracted from Japanese Callicarpa, acts as a p38-alpha MAPK inhibitor and exhibits anti-inflammatory properties. Purity 99.11%
Catalog No. 50-287-8219 Supplier TARGETMOL CHEMICALS INC Supplier No. T705125MG
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. 5,6,7-Trimethoxyflavone, a natural product extracted from Japanese Callicarpa, acts as a p38-alpha MAPK inhibitor and exhibits anti-inflammatory properties. Purity 99.11%