Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5,6-Benzo-4,7,13,16,21,24-Hexaoxa-1,10-Diazabicyclo[8.8.8]Hexacos-5-Ene Solution

Catalog No. 308579100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
30-857-9100MG 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 30-857-9100MG Supplier Sigma Aldrich Supplier No. 308579100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

5,6-Benzo-4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacos-5-ene can be used as a model ligand:

  • In the metal-ligand complex formation studies to predict ligand coordination with metal ions (Eu & Gd) using magnetic resonance imaging (MRI) technique.
  • In the study of coordination environment as well as redox and electronic properties of its YbII complexes.Kryptofix is a registered trademark of Merck KGaA, Darmstadt, Germany

Specifications

CAS 63348-81-2
Density 1.059 g/mL at 25°C
Molecular Formula C2H8BClS
Refractive Index n20/D 1.516
Linear Formula (CH3)2S-+BH2Cl
MDL Number MFCD00013210
Quantity 50 g
Synonym Kryptofix(R) 222B; Kryptand 222B
Molecular Weight (g/mol) 110.41 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.