Learn More
Medchemexpress LLC 5(6)-Carboxy-2',7'-dichlorofluorescein | 111843-78-8 | 99.0% | 100 MG
SDP
Catalog No. 5000409756
Encompass_Preferred
5(6)-Carboxy-2',7'-dichlorofluorescein is an ideal substrate for MRP2 vesicular transport assay, possessing excellent detection and transport properties.
  • Ideal substrate for MRP2 vesicular transport assay
  • Excellent detection and transport properties
Catalog No. 50-004-09756 Supplier Medchemexpress LLC Supplier No. HYD0194100MG
5(6)-Carboxy-2',7'-dichlorofluorescein is an ideal substrate for MRP2 vesicular transport assay, possessing excellent detection and transport properties.
  • Ideal substrate for MRP2 vesicular transport assay
  • Excellent detection and transport properties