Learn More
Medchemexpress LLC 5(6)-Carboxy-2',7'-dichlorofluorescein | 111843-78-8 | 99.03% | 25 MG
SDP
Catalog No. 5000411644
Encompass_Preferred
5(6)-Carboxy-2',7'-dichlorofluorescein is an ideal substrate for MRP2 vesicular transport assay, with excellent detection and transport properties.
  • Ideal substrate for MRP2 vesicular transport assay.
  • Excellent detection properties.
  • Excellent transport properties.
  • For research use only.
Catalog No. 50-004-11644 Supplier Medchemexpress LLC Supplier No. 25MGHYD0194
5(6)-Carboxy-2',7'-dichlorofluorescein is an ideal substrate for MRP2 vesicular transport assay, with excellent detection and transport properties.
  • Ideal substrate for MRP2 vesicular transport assay.
  • Excellent detection properties.
  • Excellent transport properties.
  • For research use only.