Learn More
Medchemexpress LLC 5(6)-Carboxy-2',7'-dichlorofluorescein | 111843-78-8 | 99.03% | 50 MG
SDP
Catalog No. 5000410768
Encompass_Preferred
5(6)-Carboxy-2',7'-dichlorofluorescein is an ideal substrate for MRP2 vesicular transport assay, with excellent detection and transport properties. It is for research use only.
  • Ideal substrate for MRP2 vesicular transport assay
  • Excellent detection properties
  • Excellent transport properties
Catalog No. 50-004-10768 Supplier Medchemexpress LLC Supplier No. HYD019450MG
5(6)-Carboxy-2',7'-dichlorofluorescein is an ideal substrate for MRP2 vesicular transport assay, with excellent detection and transport properties. It is for research use only.
  • Ideal substrate for MRP2 vesicular transport assay
  • Excellent detection properties
  • Excellent transport properties