Learn More
Medchemexpress LLC 5(6)-carboxy-2',7'-dichlorofluorescein diacetate | 127770-45-0 | 97.5% | 529.28 | C25H14Cl2O9 | 10 MG
SDP
Catalog No. 5000297839
Encompass_Preferred
5(6)-Carboxy-2',7'-dichlorofluorescein diacetate is a cell-permeant fluorescent tracer used to detect reactive oxygen species (ROS) in live-cell assays. Supplied as the diacetate ester to enable cellular uptake, intracellular esterases cleave the acetate groups to yield the fluorescent carboxy-dichlorofluorescein. It is commonly used in fluorescence-based assays and live-cell imaging.
Catalog No. 50-002-97839 Supplier Medchemexpress LLC Supplier No. HYD107810MG
5(6)-Carboxy-2',7'-dichlorofluorescein diacetate is a cell-permeant fluorescent tracer used to detect reactive oxygen species (ROS) in live-cell assays. Supplied as the diacetate ester to enable cellular uptake, intracellular esterases cleave the acetate groups to yield the fluorescent carboxy-dichlorofluorescein. It is commonly used in fluorescence-based assays and live-cell imaging.