Learn More
Cayman Chemical 5 6-CarboxyfluoresceIn 1g
SDP
Catalog No. 502965862
Encompass_Preferred
A mixture of 5-carboxy and 6-carboxy derivatives of fluorescein; used to label biomolecules through a reaction involving the carboxyl group; membrane impermeant, with applications in studies of membrane permeability
Catalog No. 50-296-5862 Supplier Cayman Chemical Supplier No. 171721 G
A mixture of 5-carboxy and 6-carboxy derivatives of fluorescein; used to label biomolecules through a reaction involving the carboxyl group; membrane impermeant, with applications in studies of membrane permeability