Learn More
Cayman Chemical 5 6-CarboxyfluoresceIn 5g
SDP
Catalog No. 502965868
Encompass_Preferred
A mixture of 5-carboxy and 6-carboxy derivatives of fluorescein; used to label biomolecules through a reaction involving the carboxyl group; membrane impermeant, with applications in studies of membrane permeability
Catalog No. 50-296-5868 Supplier Cayman Chemical Supplier No. 171725 G
Add to Cart
Add to Cart
A mixture of 5-carboxy and 6-carboxy derivatives of fluorescein; used to label biomolecules through a reaction involving the carboxyl group; membrane impermeant, with applications in studies of membrane permeability