Learn More
Aat Bioquest 5(6)-Carboxyrhodamine 6G cadaverine
SDP
Catalog No. 501952840
Encompass_Preferred
5(6)-Caroxyrhodamine 6G cadaverine is a building block for developing rhodamine bioconjugates. It is so a fluorescent transglutaminase substrate to label proteins by transaminidation.
Catalog No. 50-195-2840 Supplier Aat Bioquest Supplier No. 343
5(6)-Caroxyrhodamine 6G cadaverine is a building block for developing rhodamine bioconjugates. It is so a fluorescent transglutaminase substrate to label proteins by transaminidation.