Learn More
AdipoGen 5(6)-CFDA N-succinimidyl ester (50 mg)
SDP
Catalog No. 501687554
Encompass_Preferred
5(6)-CFDA N-succinimidyl ester (50 mg), Adipogen Life Sciences. Chemical. CAS: 150347-59-4. Formula: C29H19NO11. MW: 557.46. Synthetic. 5(6)-Carboxyfluorescein diacetate N-succinimidyl ester is a useful fluorescent tracer that can passivley diffuse into cells and covalently label intracellular proteins, resulting in long-term cell labeling. The reagent itself is colorless and nonfluorescent but becomes brightly green fluorescent once it is hydrolyzed by intracellular esterases.
Catalog No. 50-168-7554 Supplier AdipoGen Supplier No. CDXC0037M050
5(6)-CFDA N-succinimidyl ester (50 mg), Adipogen Life Sciences. Chemical. CAS: 150347-59-4. Formula: C29H19NO11. MW: 557.46. Synthetic. 5(6)-Carboxyfluorescein diacetate N-succinimidyl ester is a useful fluorescent tracer that can passivley diffuse into cells and covalently label intracellular proteins, resulting in long-term cell labeling. The reagent itself is colorless and nonfluorescent but becomes brightly green fluorescent once it is hydrolyzed by intracellular esterases.